Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006838.1 | NP_001006839.1 | 1 - 2 | 3034 | 107 | |
GenBank mRNA | BC076918.1 | - | 1 - 2 | 3034 | 107 | |
CR942443.2 | - | 1 - 2 | 2949 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |