Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006864.1 | NP_001006865.1 | 3 - 4 | 990 | 87 | |
XM_012956765.2 | XP_012812219.1 | 3 - 4 | 1339 | 87 | ||
XM_012956766.2 | XP_012812220.1 | 3 - 4 | 1339 | 87 | ||
XM_012956767.2 | XP_012812221.1 | 3 - 4 | 1340 | 87 | ||
GenBank mRNA | BC076974.1 | - | 3 - 4 | 990 | 87 | |
CT030354.1 | - | 3 - 4 | 921 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |