Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vezf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006864.1 | NP_001006865.1 | 4 - 5 | 1179 | 88 | |
XM_012956766.2 | XP_012812220.1 | 4 - 5 | 1528 | 88 | ||
GenBank mRNA | BC076974.1 | - | 4 - 5 | 1179 | 88 | |
CT030354.1 | - | 4 - 5 | 1110 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |