Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bcl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006871.1 | NP_001006872.1 | 1 - 2 | 160 | 95 | |
GenBank mRNA | BC076987.1 | - | 1 - 2 | 160 | 95 | |
CR926398.1 | - | 1 - 2 | 101 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |