Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006878.1 | NP_001006879.1 | 2 - 3 | 281 | 109 | |
GenBank mRNA | BC077024.1 | - | 2 - 3 | 281 | 109 | |
CR761328.2 | - | 2 - 3 | 250 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |