Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxmp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006884.2 | NP_001006885.1 | 1 - 2 | 157 | 82 | |
GenBank mRNA | BC077678.1 | - | 1 - 2 | 158 | 82 | |
CT030300.1 | - | 1 - 2 | 150 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |