Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006885.1 | NP_001006886.1 | 1 - 2 | 108 | 76 | |
XM_018097712.1 | XP_017953201.1 | 1 - 2 | 193 | 76 | ||
GenBank mRNA | BC077682.1 | - | 1 - 2 | 108 | 76 | |
CR855493.2 | - | 1 - 2 | 101 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |