Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fam3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007502.1 | NP_001007503.1 | 4 - 5 | 296 | 94 | |
XM_012960902.2 | XP_012816356.1 | 4 - 5 | 461 | 94 | ||
XM_018089361.1 | XP_017944850.1 | 4 - 5 | 316 | 94 | ||
GenBank mRNA | BC079939.1 | - | 4 - 5 | 296 | 94 | |
CR760189.2 | - | 4 - 5 | 293 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |