Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007859.1 | NP_001007860.1 | 2 - 3 | 443 | 72 | |
GenBank mRNA | BC079961.1 | - | 2 - 3 | 443 | 72 | |
CR761927.2 | - | 2 - 3 | 267 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |