Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007868.1 | NP_001007869.1 | 2 - 3 | 242 | 97 | |
XM_018097089.1 | XP_017952578.1 | 2 - 3 | 264 | 97 | ||
GenBank mRNA | BC061291.1 | - | 2 - 3 | 242 | 96 | |
BC080133.1 | - | 2 - 3 | 242 | 97 | ||
CR855816.2 | - | 2 - 3 | 240 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |