Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mettl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007898.1 | NP_001007899.1 | 2 - 3 | 449 | 88 | |
GenBank mRNA | BC080334.1 | - | 2 - 3 | 449 | 88 | |
CR760379.1 | - | 2 - 3 | 411 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |