Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ift20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007901.1 | NP_001007902.1 | 2 - 3 | 411 | 93 | |
XM_012956808.2 | XP_012812262.1 | 2 - 3 | 632 | 93 | ||
GenBank mRNA | BC080338.1 | - | 2 - 3 | 411 | 93 | |
CR762302.2 | - | 2 - 3 | 152 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |