Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cpsf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007932.1 | NP_001007933.1 | 4 - 5 | 641 | 98 | |
XM_012970321.2 | XP_012825775.1 | 3 - 4 | 384 | 98 | ||
XM_012970322.2 | XP_012825776.1 | 3 - 4 | 449 | 98 | ||
GenBank mRNA | BC080440.1 | - | 4 - 5 | 641 | 98 | |
CR761660.2 | - | 3 - 4 | 325 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |