Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ints14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007938.1 | NP_001007939.1 | 6 - 7 | 894 | 83 | |
XM_012958881.2 | XP_012814335.1 | 6 - 7 | 1328 | 83 | ||
GenBank mRNA | BC080444.1 | - | 6 - 7 | 894 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |