Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007968.2 | NP_001007969.2 | 2 - 3 | 362 | 92 | |
GenBank mRNA | BC080480.1 | - | 2 - 3 | 363 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |