Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007990.1 | NP_001007991.1 | 8 - 9 | 734 | 102 | |
XM_012953584.2 | XP_012809038.1 | 8 - 9 | 822 | 102 | ||
XM_012953585.2 | XP_012809039.1 | 8 - 9 | 833 | 102 | ||
GenBank mRNA | BC080503.1 | - | 8 - 9 | 734 | 102 | |
CR926422.2 | - | 7 - 8 | 674 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |