Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007999.1 | NP_001008000.1 | 5 - 6 | 643 | 101 | |
GenBank mRNA | BC080878.1 | - | 5 - 6 | 643 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |