Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nceh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008008.1 | NP_001008009.1 | 419 | 106 | ||
XM_012962912.2 | XP_012818366.1 | 3 - 4 | 569 | 106 | ||
XM_012962913.2 | XP_012818367.1 | 3 - 4 | 683 | 106 | ||
XM_018093851.1 | XP_017949340.1 | 3 - 4 | 394 | 106 | ||
GenBank mRNA | BC080887.1 | - | 419 | 106 | ||
CT485686.1 | - | 2 - 3 | 394 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |