Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdf2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008032.1 | NP_001008033.1 | 1 - 2 | 201 | 64 | |
GenBank mRNA | BC080914.1 | - | 1 - 2 | 201 | 64 | |
CR761046.2 | - | 1 - 2 | 179 | 64 |
Gene Symbol: LOC101732749 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004920857.2 | XP_004920914.1 | 1 - 2 | 423 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |