Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kras | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008033.1 | NP_001008034.1 | 3 - 4 | 500 | 72 | |
GenBank mRNA | BC080916.1 | - | 3 - 4 | 500 | 72 | |
CR760764.2 | - | 3 - 4 | 500 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |