Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdelr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008051.1 | NP_001008052.1 | 2 - 3 | 514 | 109 | |
GenBank mRNA | BC080938.1 | - | 2 - 3 | 514 | 109 | |
CR942706.2 | - | 2 - 3 | 216 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |