Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dlx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008060.1 | NP_001008061.1 | 2 - 3 | 691 | 103 | |
GenBank mRNA | BC080947.1 | - | 2 - 3 | 691 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |