Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008074.1 | NP_001008075.1 | 5 - 6 | 681 | 61 | |
GenBank mRNA | BC080969.1 | - | 5 - 6 | 681 | 61 | |
CR760859.2 | - | 5 - 6 | 678 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |