Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: m6pr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008114.1 | NP_001008115.1 | 4 - 5 | 676 | 68 | |
XM_012956817.2 | XP_012812271.1 | 5 - 6 | 654 | 68 | ||
XM_012956818.2 | XP_012812272.1 | 4 - 5 | 546 | 68 | ||
GenBank mRNA | BC081319.1 | - | 4 - 5 | 676 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |