Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008129.1 | NP_001008130.1 | 4 - 5 | 476 | 80 | |
XM_012971685.2 | XP_012827139.1 | 4 - 5 | 564 | 80 | ||
XM_012971686.2 | XP_012827140.1 | 4 - 5 | 545 | 80 | ||
XM_012971687.2 | XP_012827141.1 | 4 - 5 | 480 | 80 | ||
XM_012971688.2 | XP_012827142.1 | 4 - 5 | 480 | 80 | ||
XM_012971689.2 | XP_012827143.1 | 4 - 5 | 564 | 80 | ||
GenBank mRNA | BC081338.1 | - | 4 - 5 | 476 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |