Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008134.2 | NP_001008135.2 | 4 - 5 | 798 | 69 | |
XM_018096228.1 | XP_017951717.1 | 4 - 5 | 789 | 69 | ||
GenBank mRNA | BC081343.1 | - | 4 - 5 | 715 | 69 | |
CR942386.2 | - | 4 - 5 | 706 | 69 | ||
CX329479.2 | - | 4 - 5 | 798 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |