Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008150.1 | NP_001008151.1 | 6 - 7 | 517 | 83 | |
XM_012971690.2 | XP_012827144.1 | 7 - 8 | 606 | 83 | ||
XM_012971691.2 | XP_012827145.1 | 6 - 7 | 577 | 83 | ||
XM_012971692.2 | XP_012827146.1 | 5 - 6 | 515 | 83 | ||
XM_012971693.2 | XP_012827147.1 | 4 - 5 | 457 | 83 | ||
XM_018097717.1 | XP_017953206.1 | 6 - 7 | 544 | 83 | ||
GenBank mRNA | BC081364.1 | - | 6 - 7 | 517 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |