Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: strada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008150.1 | NP_001008151.1 | 9 - 10 | 918 | 64 | |
XM_012971690.2 | XP_012827144.1 | 10 - 11 | 1007 | 64 | ||
XM_012971691.2 | XP_012827145.1 | 9 - 10 | 978 | 64 | ||
XM_012971692.2 | XP_012827146.1 | 8 - 9 | 916 | 64 | ||
XM_012971693.2 | XP_012827147.1 | 7 - 8 | 858 | 64 | ||
XM_018097717.1 | XP_017953206.1 | 9 - 10 | 945 | 64 | ||
GenBank mRNA | BC081364.1 | - | 9 - 10 | 918 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |