Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008162.1 | NP_001008163.1 | 4 - 5 | 668 | 88 | |
XM_012966481.2 | XP_012821935.1 | 4 - 5 | 1428 | 88 | ||
GenBank mRNA | BC082344.1 | - | 4 - 5 | 668 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |