Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdhaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008178.1 | NP_001008179.1 | 1 - 2 | 173 | 60 | |
GenBank mRNA | BC082498.1 | - | 1 - 2 | 173 | 60 | |
CR761009.2 | - | 1 - 2 | 133 | 60 | ||
CR926240.1 | - | 1 - 2 | 133 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |