Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: utp14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008204.1 | NP_001008205.1 | 8 - 9 | 841 | 82 | |
XM_012968277.2 | XP_012823731.1 | 8 - 9 | 817 | 82 | ||
XM_012968278.2 | XP_012823732.1 | 8 - 9 | 898 | 82 | ||
GenBank mRNA | BC080963.1 | - | 8 - 9 | 841 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |