Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: max | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008207.1 | NP_001008208.1 | 2 - 3 | 287 | 103 | |
XM_012970352.2 | XP_012825806.1 | 3 - 4 | 978 | 103 | ||
GenBank mRNA | BC081313.1 | - | 2 - 3 | 287 | 103 | |
CR942797.2 | - | 3 - 4 | 441 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |