Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008430.3 | NP_001008430.2 | 2 - 3 | 231 | 114 | |
XM_012962908.2 | XP_012818362.1 | 2 - 3 | 415 | 114 | ||
GenBank mRNA | AL779600.2 | - | 2 - 3 | 232 | 114 | |
BC080134.1 | - | 2 - 3 | 220 | 114 | ||
CF240668.1 | - | 2 - 3 | 220 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |