Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008439.1 | NP_001008439.1 | 4 - 5 | 465 | 71 | |
GenBank mRNA | BC059755.1 | - | 4 - 5 | 439 | 71 | |
BC080337.1 | - | 4 - 5 | 465 | 71 | ||
CR761691.2 | - | 4 - 5 | 422 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |