Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pa2g4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008439.1 | NP_001008439.1 | 8 - 9 | 783 | 96 | |
GenBank mRNA | BC059755.1 | - | 8 - 9 | 757 | 96 | |
BC080337.1 | - | 8 - 9 | 783 | 96 | ||
CR761691.2 | - | 8 - 9 | 740 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |