Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011102.1 | NP_001011102.1 | 2 - 3 | 553 | 97 | |
GenBank mRNA | BC084499.1 | - | 2 - 3 | 553 | 97 | |
CR855466.2 | - | 1 - 2 | 482 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |