Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011102.1 | NP_001011102.1 | 4 - 5 | 784 | 78 | |
GenBank mRNA | BC084499.1 | - | 4 - 5 | 784 | 78 | |
CR855466.2 | - | 3 - 4 | 713 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |