Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnni2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011110.2 | NP_001011110.1 | 4 - 5 | 126 | 60 | |
XM_012960980.1 | XP_012816434.1 | 4 - 5 | 173 | 60 | ||
XM_012960981.1 | XP_012816435.1 | 3 - 4 | 547 | 60 | ||
GenBank mRNA | BC084508.1 | - | 4 - 5 | 127 | 60 | |
CU464111.1 | - | 4 - 5 | 103 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Xenopus tropicalis Probe spans exons |