Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011121.2 | NP_001011121.1 | 2 - 3 | 198 | 101 | |
GenBank mRNA | BC084520.1 | - | 2 - 3 | 199 | 101 | |
CT025369.2 | - | 2 - 3 | 184 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |