Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011139.1 | NP_001011139.1 | 4 - 5 | 523 | 75 | |
GenBank mRNA | BC084468.1 | - | 4 - 5 | 523 | 75 | |
CR855769.2 | - | 4 - 5 | 522 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |