Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psmg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011145.1 | NP_001011145.1 | 5 - 6 | 612 | 69 | |
GenBank mRNA | BC084517.1 | - | 5 - 6 | 612 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |