Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prss27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018097584.1 | XP_017953073.1 | 2 - 3 | 317 | 93 | |
GenBank mRNA | BC087611.1 | - | 2 - 3 | 80 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |