Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cplx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011205.1 | NP_001011205.1 | 11 - 12 | 1290 | 99 | |
GenBank mRNA | BC087752.1 | - | 11 - 12 | 1290 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |