Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cplx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011205.1 | NP_001011205.1 | 12 - 13 | 1414 | 70 | |
GenBank mRNA | BC087752.1 | - | 12 - 13 | 1414 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |