Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: try10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011209.1 | NP_001011209.1 | 3 - 4 | 465 | 74 | |
GenBank mRNA | BC087759.1 | - | 3 - 4 | 465 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |