Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011215.1 | NP_001011215.1 | 3 - 4 | 445 | 86 | |
GenBank mRNA | BC087771.1 | - | 3 - 4 | 445 | 86 | |
CR761228.2 | - | 3 - 4 | 460 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |