Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011219.1 | NP_001011219.1 | 5 - 6 | 563 | 67 | |
GenBank mRNA | BC087779.1 | - | 5 - 6 | 563 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |