Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011225.1 | NP_001011225.1 | 8 - 9 | 728 | 81 | |
XM_012960232.1 | XP_012815686.1 | 10 - 11 | 973 | 81 | ||
XM_012960233.1 | XP_012815687.1 | 10 - 11 | 958 | 81 | ||
XM_012960234.2 | XP_012815688.1 | 10 - 11 | 969 | 81 | ||
XM_012960235.1 | XP_012815689.1 | 8 - 9 | 820 | 81 | ||
XM_012960236.2 | XP_012815690.1 | 7 - 8 | 645 | 81 | ||
GenBank mRNA | BC087789.1 | - | 8 - 9 | 728 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |