Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elovl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011248.1 | NP_001011248.1 | 3 - 4 | 318 | 92 | |
XM_018093868.1 | XP_017949357.1 | 3 - 4 | 662 | 92 | ||
GenBank mRNA | BC087826.1 | - | 3 - 4 | 318 | 92 | |
CT030337.1 | - | 3 - 4 | 316 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |