Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vdac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011249.2 | NP_001011249.1 | 2 - 3 | 123 | 62 | |
XM_012958926.2 | XP_012814380.1 | 3 - 4 | 219 | 62 | ||
XM_012958931.1 | XP_012814385.1 | 3 - 4 | 199 | 62 | ||
XM_012958932.2 | XP_012814386.1 | 3 - 4 | 203 | 62 | ||
GenBank mRNA | BC087828.1 | - | 2 - 3 | 124 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |